Learn More
Trajan Scientific Column BP20 QC1 Medical 1/EA
SDP
Catalog No. 5000494502
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length BP20 GC capillary column
Catalog No. 50-004-94502 Supplier Trajan Scientific Supplier No. 054405
0 1 mm ID x 0 1 m film thickness x 10 m length BP20 GC capillary column