Learn More
Trajan Scientific Column BP20 QC2.5 Medical 1/EA
SDP
Catalog No. 5000493620
Encompass_Preferred
0 25 mm ID x 0 5 m film thickness x 30 m length BP20 GC capillary column
This item is not returnable. View return policy
Catalog No. 50-004-93620 Supplier Trajan Scientific Supplier No. 054415
0 25 mm ID x 0 5 m film thickness x 30 m length BP20 GC capillary column