Learn More
Trajan Scientific Column BP20 QC2.5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054427
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length BP20 GC capillary colum
Catalog No. 50-004-94253