Learn More
Trajan Scientific Column BP20 QC2 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054424
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 30 m length BP20 GC capillary colum
Catalog No. 50-004-93608