Learn More
Trajan Scientific Column BP20 QC2 Medical 1/EA
SDP
Catalog No. 5000494460
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 50 m length BP20 GC capillary colum
Catalog No. 50-004-94460 Supplier Trajan Scientific Supplier No. 054422
0 22 mm ID x 0 25 m film thickness x 50 m length BP20 GC capillary colum