Learn More
Trajan Scientific Column BP20 QC3 5 in 1/EA
SDP
Catalog No. 5000493764
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 60 m length BP20 GC capillary column on 5 cage
Catalog No. 50-004-93764 Supplier Trajan Scientific Supplier No. 054457CCS
0 32 mm ID x 0 5 m film thickness x 60 m length BP20 GC capillary column on 5 cage