Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054434
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 60 m length BP20 GC capillary colum
Catalog No. 50-004-94193