Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Catalog No. 5000494140
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length BP20 GC capillary colum
Catalog No. 50-004-94140 Supplier Trajan Scientific Supplier No. 054433
0 32 mm ID x 0 25 m film thickness x 30 m length BP20 GC capillary colum