Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Catalog No. 5000493746
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 50 m length BP20 GC capillary colum
Catalog No. 50-004-93746 Supplier Trajan Scientific Supplier No. 054431
0 32 mm ID x 0 25 m film thickness x 50 m length BP20 GC capillary colum