Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Catalog No. 5000493566
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 50 m length BP20 GC capillary column
Catalog No. 50-004-93566 Supplier Trajan Scientific Supplier No. 054443
Add to Cart
Add to Cart
0 32 mm ID x 1 0 m film thickness x 50 m length BP20 GC capillary column