Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054442
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 25 m length BP20 GC capillary column
Catalog No. 50-004-93610