Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Catalog No. 5000494878
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 30 m length BP20 GC capillary column
Catalog No. 50-004-94878 Supplier Trajan Scientific Supplier No. 054444
0 32 mm ID x 1 0 m film thickness x 30 m length BP20 GC capillary column