Learn More
Trajan Scientific Column BP20 QC3 Medical 1/EA
SDP
Catalog No. 5000493619
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 60 m length BP20 GC capillary column
Catalog No. 50-004-93619 Supplier Trajan Scientific Supplier No. 054457
0 32 mm ID x 0 5 m film thickness x 60 m length BP20 GC capillary column