Learn More
Trajan Scientific Column BP20 QC5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054440
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 30 m length BP20 GC capillary column
Catalog No. 50-004-93632