Learn More
Trajan Scientific Column BP20 QC5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054456
Encompass_Preferred
0 53 mm ID x 2 0 m film thickness x 25 m length BP20 GC capillary column
Catalog No. 50-004-93646