Learn More
Trajan Scientific Column BP20 QC5 Medical 1/EA
SDP
Catalog No. 5000493881
Encompass_Preferred
0 53 mm ID x 1 0 m film thickness x 25 m length BP20 GC capillary column
Catalog No. 50-004-93881 Supplier Trajan Scientific Supplier No. 054448
0 53 mm ID x 1 0 m film thickness x 25 m length BP20 GC capillary column