Learn More
Trajan Scientific Column BP21 QC2.5 Medical 1/EA
SDP
Catalog No. 5000494282
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 60 m length BP21 GC capillary colum
Catalog No. 50-004-94282 Supplier Trajan Scientific Supplier No. 054466
0 25 mm ID x 0 25 m film thickness x 60 m length BP21 GC capillary colum