Learn More
Trajan Scientific Column BP21 QC2 Medical 1/EA
SDP
Catalog No. 5000494530
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 25 m length BP21 GC capillary colum
Catalog No. 50-004-94530 Supplier Trajan Scientific Supplier No. 054462
0 22 mm ID x 0 25 m film thickness x 25 m length BP21 GC capillary colum