Learn More
Trajan Scientific Column BP21 QC3 Medical 1/EA
SDP
Catalog No. 5000493609
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 60 m length BP21 GC capillary colum
Catalog No. 50-004-93609 Supplier Trajan Scientific Supplier No. 054472
0 32 mm ID x 0 25 m film thickness x 60 m length BP21 GC capillary colum