Learn More
Trajan Scientific Column BP21 QC5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054474
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 25 m length BP21 GC capillary column
Catalog No. 50-004-94618