Learn More
Trajan Scientific Column BP21 QC5 Medical 1/EA
SDP
Catalog No. 5000493821
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 30 m length BP21 GC capillary column
Catalog No. 50-004-93821 Supplier Trajan Scientific Supplier No. 054477
Add to Cart
Add to Cart
0 53 mm ID x 0 5 m film thickness x 30 m length BP21 GC capillary column