Learn More
Trajan Scientific Column BP21 QC5 Medical 1/EA
SDP
Catalog No. 5000493633
Encompass_Preferred
0 53 mm ID x 1 0 m film thickness x 30 m length BP21 GC capillary column
Catalog No. 50-004-93633 Supplier Trajan Scientific Supplier No. 054478
0 53 mm ID x 1 0 m film thickness x 30 m length BP21 GC capillary column