Learn More
Trajan Scientific Column BP5 QC2.5 5 in 1/EA
SDP
Catalog No. 5000493770
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length BP5 GC capillary column on 5 cage
Catalog No. 50-004-93770 Supplier Trajan Scientific Supplier No. 054183CCS
0 25 mm ID x 0 25 m film thickness x 30 m length BP5 GC capillary column on 5 cage