Learn More
Trajan Scientific Column BP5 QC2 Medical 1/EA
SDP
Catalog No. 5000494379
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 12 m length BP5 GC capillary column
Catalog No. 50-004-94379 Supplier Trajan Scientific Supplier No. 054167
0 22 mm ID x 0 25 m film thickness x 12 m length BP5 GC capillary column