Learn More
Trajan Scientific Column BP5 QC3 Medical 1/EA
SDP
Catalog No. 5000494620
Encompass_Preferred
0 32 mm ID x 1 0 m film thickness x 30 m length BP5 GC capillary column
Catalog No. 50-004-94620 Supplier Trajan Scientific Supplier No. 054189
0 32 mm ID x 1 0 m film thickness x 30 m length BP5 GC capillary column