Learn More
Trajan Scientific Column BP5 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054186
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 25 m length BP5 GC capillary column
Catalog No. 50-004-94270