Learn More
Trajan Scientific Column BP5 QC5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054199
Encompass_Preferred
0 53 mm ID x 1 5 m film thickness x 15 m length BP5 GC capillary column
Catalog No. 50-004-93653