Learn More
Trajan Scientific Column BP5MS QC2.5 Medical 1/EA
SDP
Catalog No. 5000494673
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 15 m length BP5MS GC capillary column
Catalog No. 50-004-94673 Supplier Trajan Scientific Supplier No. 054308
Add to Cart
Add to Cart
0 25 mm ID x 0 25 m film thickness x 15 m length BP5MS GC capillary column