Learn More
Trajan Scientific Column BP5MS QC2.5 SilGuard 1/EA
SDP
Catalog No. 5000493984
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length BP5MS GC capillary column with 0 25 mm ID SilGuard
Catalog No. 50-004-93984 Supplier Trajan Scientific Supplier No. 054310SG25
0 25 mm ID x 0 25 m film thickness x 30 m length BP5MS GC capillary column with 0 25 mm ID SilGuard