Learn More
Trajan Scientific Column BP624 QC3 5 in 1/EA
SDP
Catalog No. 5000494674
Encompass_Preferred
0 32 mm ID x 1 8 m film thickness x 30 m length BP624 GC capillary column on 5 cage
Catalog No. 50-004-94674 Supplier Trajan Scientific Supplier No. 054832CCS
0 32 mm ID x 1 8 m film thickness x 30 m length BP624 GC capillary column on 5 cage