Learn More
Trajan Scientific Column BP624 QC5 Medical 1/EA
SDP
Catalog No. 5000493684
Encompass_Preferred
0 53 mm ID x 3 0 m film thickness x 30 m length BP624 GC capillary column
Catalog No. 50-004-93684 Supplier Trajan Scientific Supplier No. 054836
0 53 mm ID x 3 0 m film thickness x 30 m length BP624 GC capillary column