Learn More
Trajan Scientific Column BP624 QC5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054835
Encompass_Preferred
0 53 mm ID x 3 0 m film thickness x 50 m length BP624 GC capillary column
Catalog No. 50-004-93832