Learn More
Trajan Scientific Column BPX BIODIESEL QC3 6584 1/EA
SDP
Catalog No. 5000493840
Encompass_Preferred
0 32 mm ID x 0 1 m film thickness x 10 m length BPX-BIOD6584 with guard column
Catalog No. 50-004-93840 Supplier Trajan Scientific Supplier No. 0541180SG53
0 32 mm ID x 0 1 m film thickness x 10 m length BPX-BIOD6584 with guard column