Learn More
Trajan Scientific Column BPX DXN QC2.5 DIOXIN 1/EA
SDP
Catalog No. 5000494249
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 60 m length BPX-Dioxin GC capillary column
Catalog No. 50-004-94249 Supplier Trajan Scientific Supplier No. 054235
0 25 mm ID x 0 25 m film thickness x 60 m length BPX-Dioxin GC capillary column