Learn More
Trajan Scientific Column BPX VOLATILES QC2.5 Volatiles 1/EA
SDP
Catalog No. 5000494100
Encompass_Preferred
0 25 mm ID x 1 4 m film thickness x 30 m length BPX-Volatiles GC capillary column
Catalog No. 50-004-94100 Supplier Trajan Scientific Supplier No. 054980
0 25 mm ID x 1 4 m film thickness x 30 m length BPX-Volatiles GC capillary column