Learn More
Trajan Scientific Column BPX VOLATILES QC3 Volatiles 1/EA
SDP
Catalog No. 5000494316
Encompass_Preferred
0 32 mm ID x 1 8 m film thickness x 30 m length BPX-Volatiles GC capillary column
Catalog No. 50-004-94316 Supplier Trajan Scientific Supplier No. 054982
0 32 mm ID x 1 8 m film thickness x 30 m length BPX-Volatiles GC capillary column