Learn More
Trajan Scientific Column BPX VOLATILES QC5 Volatiles 1/EA
SDP
Catalog No. 5000493959
Encompass_Preferred
0 53 mm ID x 3 0 m film thickness x 30 m length BPX-Volatiles GC capillary column
Catalog No. 50-004-93959 Supplier Trajan Scientific Supplier No. 054984
0 53 mm ID x 3 0 m film thickness x 30 m length BPX-Volatiles GC capillary column