Learn More
Trajan Scientific Column BPX VOLATILES QC5 Volatiles 1/EA
SDP
Supplier:  Trajan Scientific 054985
Encompass_Preferred
0 53 mm ID x 3 0 m film thickness x 60 m length BPX-Volatiles GC capillary column
Catalog No. 50-004-94538