Learn More
Trajan Scientific Column BPX1 QC1 Fast 1/EA
SDP
Catalog No. 5000493913
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length BPX1 GC capillary col mn
Catalog No. 50-004-93913 Supplier Trajan Scientific Supplier No. 054777
0 1 mm ID x 0 1 m film thickness x 10 m length BPX1 GC capillary col mn