Learn More
Trajan Scientific Column BPX35 QC1 Medical 1/EA
SDP
Catalog No. 5000494272
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length BPX35 GC capillary column
Catalog No. 50-004-94272 Supplier Trajan Scientific Supplier No. 054699
0 1 mm ID x 0 1 m film thickness x 10 m length BPX35 GC capillary column