Learn More
Trajan Scientific Column BPX35 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 0547158
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 30 m length BPX35 GC capillary column
Catalog No. 50-004-93659