Learn More
Trajan Scientific Column BPX35 QC3 Medical 1/EA
SDP
Catalog No. 5000494356
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length BPX35 GC capillary column
Catalog No. 50-004-94356 Supplier Trajan Scientific Supplier No. 054724
0 32 mm ID x 0 25 m film thickness x 30 m length BPX35 GC capillary column