Learn More
Trajan Scientific Column BPX35 QC5 5 in 1/EA
SDP
Catalog No. 5000494251
Encompass_Preferred
0 53 mm ID x 1 0 m film thickness x 30 m length BPX35 GC capillary column 5 cage
Catalog No. 50-004-94251 Supplier Trajan Scientific Supplier No. 054737CCS
0 53 mm ID x 1 0 m film thickness x 30 m length BPX35 GC capillary column 5 cage