Learn More
Trajan Scientific Column BPX5 QC1.5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054110
Encompass_Preferred
0 15 mm ID x 0 15 m film thickness x 30 m length BPX5 GC capillary column
Catalog No. 50-004-94879