Learn More
Trajan Scientific Column BPX5 QC1 Medical 1/EA
SDP
Catalog No. 5000493956
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length BPX5 GC capillary column
Catalog No. 50-004-93956 Supplier Trajan Scientific Supplier No. 054099
Add to Cart
Add to Cart
0 1 mm ID x 0 1 m film thickness x 10 m length BPX5 GC capillary column