Learn More
Trajan Scientific Column BPX5 QC2.5 5 in 1/EA
SDP
Catalog No. 5000493769
Encompass_Preferred
0 25 mm ID x 0 1 m film thickness x 30 m length BPX5 GC capillary column on 5 cage
Catalog No. 50-004-93769 Supplier Trajan Scientific Supplier No. 054101CCS
0 25 mm ID x 0 1 m film thickness x 30 m length BPX5 GC capillary column on 5 cage