Learn More
Trajan Scientific Column BPX5 QC2.5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 0541011
Encompass_Preferred
0 25 mm ID x 0 1 m film thickness x 30 m length BPX5 GC capillary column
Catalog No. 50-004-93657