Learn More
Trajan Scientific Column BPX5 QC2.5 Medical 1/EA
SDP
Catalog No. 5000493616
Encompass_Preferred
0 25 mm ID x 0 1 m film thickness x 15 m length BPX5 GC capillary column
Catalog No. 50-004-93616 Supplier Trajan Scientific Supplier No. 0542170
0 25 mm ID x 0 1 m film thickness x 15 m length BPX5 GC capillary column