Learn More
Trajan Scientific Column BPX5 QC2.5 SilGuard 1/EA
SDP
Catalog No. 5000494687
Encompass_Preferred
0 25 mm ID x 0 1 m film thickness x 30 m length BPX5 GC capillary column with 0 32 mm ID SilGuard
Catalog No. 50-004-94687 Supplier Trajan Scientific Supplier No. 054101SG32
0 25 mm ID x 0 1 m film thickness x 30 m length BPX5 GC capillary column with 0 32 mm ID SilGuard