Learn More
Trajan Scientific Column BPX5 QC2 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054112
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 12 m length BPX5 GC capillary column
Catalog No. 50-004-94124